Assay Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745868.3 | XP_002745914.1 | 4 - 5 | 605 | 105 | |
| XM_008993387.1 | XP_008991635.1 | 4 - 5 | 599 | 105 | ||
| XM_008993388.2 | XP_008991636.1 | 4 - 5 | 554 | 105 | ||
More Information
Set Membership: |
|

COMMD8
Genomic Map