Assay Details
| Gene Symbol: TNFRSF10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979187.2 | XP_008977435.2 | 8 - 9 | 1016 | 84 | |
| XM_017963691.1 | XP_017819180.1 | 7 - 8 | 1103 | 84 | ||
More Information
Set Membership: |
|

TNFRSF10B
Genomic Map