Assay Details
| Gene Symbol: LLGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806786.4 | XP_002806832.2 | 22 - 23 | 3264 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LLGL1| Gene Symbol: LLGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806786.4 | XP_002806832.2 | 22 - 23 | 3264 | 56 | |
Set Membership: |
|