Assay Details
| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004888.2 | XP_009003136.1 | 703 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RILPL2| Gene Symbol: RILPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004888.2 | XP_009003136.1 | 703 | 96 | ||
Set Membership: |
|