Assay Details
| Gene Symbol: ITPKC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762140.3 | XP_002762186.1 | 2 - 3 | 1389 | 72 | |
| XM_017967462.1 | XP_017822951.1 | 2 - 3 | 1389 | 72 | ||
More Information
Set Membership: |
|

ITPKC
Genomic Map