Assay Details
| Gene Symbol: C22H19orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987229.1 | XP_008985477.1 | 3 - 4 | 498 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22H19orf66| Gene Symbol: C22H19orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987229.1 | XP_008985477.1 | 3 - 4 | 498 | 86 | |