Assay Details
| Gene Symbol: ACKR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984690.2 | XP_008982938.1 | 2 - 3 | 217 | 86 | |
| XM_008984691.2 | XP_008982939.1 | 2 - 3 | 212 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008984690.2, XM_008984691.2 transcripts.
Set Membership: |
|

ACKR1
Genomic Map