Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984482.2 | XP_008982730.1 | 9 - 10 | 671 | 105 | |
| XM_008984483.2 | XP_008982731.1 | 8 - 9 | 604 | 105 | ||
| XM_008984487.2 | XP_008982735.1 | 7 - 8 | 443 | 105 | ||
| XM_017966309.1 | XP_017821798.1 | 10 - 11 | 854 | 105 | ||
| XM_017966310.1 | XP_017821799.1 | 9 - 10 | 795 | 105 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map