Assay Details
| Gene Symbol: SPTLC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747453.3 | XP_002747499.1 | 4 - 5 | 980 | 57 | |
| XM_008995768.2 | XP_008994016.1 | 5 - 6 | 852 | 57 | ||
More Information
Set Membership: |
|

SPTLC3
Genomic Map