Assay Details
Gene Symbol: CD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752244.4 | XP_002752290.2 | 7 - 8 | 798 | 66 | |
GenBank mRNA | EF215132.1 | - | 3 - 4 | 255 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
