Assay Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982830.2 | XP_008981078.1 | 1 - 2 | 594 | 69 | |
| XM_017966511.1 | XP_017822000.1 | 1 - 2 | 594 | 69 | ||
More Information
Set Membership: |
|

TRIM4
Genomic Map