Assay Details
| Gene Symbol: ATOH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980392.1 | XP_008978640.1 | 1 - 2 | 1174 | 63 | |
| XM_008980393.1 | XP_008978641.1 | 1 - 2 | 1174 | 63 | ||
| XM_017964192.1 | XP_017819681.1 | 1 - 2 | 1174 | 63 | ||
More Information
Set Membership: |
|

ATOH8
Genomic Map