Assay Details
| Gene Symbol: POLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748152.3 | XP_002748198.1 | 3 - 4 | 3109 | 80 | |
| XM_008996917.2 | XP_008995165.1 | 3 - 4 | 3109 | 80 | ||
| XM_008996918.2 | XP_008995166.1 | 3 - 4 | 3109 | 80 | ||
More Information
Set Membership: |
|

POLG2
Genomic Map