Assay Details
| Gene Symbol: MOB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757601.4 | XP_002757647.1 | 5 - 6 | 861 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOB1A| Gene Symbol: MOB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757601.4 | XP_002757647.1 | 5 - 6 | 861 | 72 | |
Set Membership: |
|