Assay Details
| Gene Symbol: GGTA1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267732.1 | NP_001254661.1 | 1 - 2 | 89 | 103 | |
| XM_009001954.2 | XP_009000202.1 | 3 - 4 | 401 | 103 | ||
| XM_017967738.1 | XP_017823227.1 | 3 - 4 | 299 | 103 | ||
| XM_017969840.1 | XP_017825329.1 | 3 - 4 | 299 | 103 | ||
| GenBank mRNA | AF384428.1 | - | 1 - 2 | 89 | 103 | |
More Information
Set Membership: |
|

GGTA1P
Genomic Map