Assay Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000346.1 | XP_008998594.1 | 3 - 4 | 1036 | 72 | |
| GenBank mRNA | EF215186.1 | - | 1 - 2 | 80 | 72 | |
More Information
Set Membership: |
|

PHF13
Genomic Map