Assay Details
| Gene Symbol: HSD17B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752619.4 | XP_002752665.1 | 4 - 5 | 866 | 69 | |
| XM_009004042.2 | XP_009002290.1 | 5 - 6 | 1084 | 69 | ||
More Information
Set Membership: |
|

HSD17B6
Genomic Map