Assay Details
| Gene Symbol: RASSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978418.1 | XP_017833907.1 | 1 - 2 | 519 | 71 | |
| XM_017978419.1 | XP_017833908.1 | 1 - 2 | 519 | 71 | ||
| XM_017978420.1 | XP_017833909.1 | 1 - 2 | 519 | 71 | ||
More Information
Set Membership: |
|

RASSF7
Genomic Map