Assay Details
| Gene Symbol: ATPAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001168.2 | XP_008999416.1 | 1 - 2 | 937 | 63 | |
| XM_009001169.2 | XP_008999417.1 | 1 - 2 | 937 | 63 | ||
More Information
Set Membership: |
|

ATPAF1
Genomic Map