Assay Details
| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757351.3 | XP_002757397.3 | 2 - 3 | 465 | 96 | |
| XM_017963573.1 | XP_017819062.1 | 2 - 3 | 199 | 96 | ||
| XM_017963574.1 | XP_017819063.1 | 2 - 3 | 201 | 96 | ||
More Information
Set Membership: |
|

TXNL4A
Genomic Map