Assay Details
| Gene Symbol: ITM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763035.4 | XP_002763081.1 | 4 - 5 | 927 | 105 | |
| XM_002763036.4 | XP_002763082.1 | 3 - 4 | 756 | 105 | ||
More Information
Set Membership: |
|

ITM2A
Genomic Map