Assay Details
| Gene Symbol: ALG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758125.4 | XP_002758171.1 | 5 - 6 | 782 | 74 | |
| XM_008981316.1 | XP_008979564.1 | 5 - 6 | 782 | 74 | ||
| XM_008981318.1 | XP_008979566.1 | 5 - 6 | 717 | 74 | ||
| XM_008981319.2 | XP_008979567.1 | 5 - 6 | 720 | 74 | ||
| XM_017964718.1 | XP_017820207.1 | 5 - 6 | 699 | 74 | ||
More Information
Set Membership: |
|

ALG3
Genomic Map