Assay Details
| Gene Symbol: NIPAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965669.1 | XP_017821158.1 | 1 - 2 | 395 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NIPAL2| Gene Symbol: NIPAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965669.1 | XP_017821158.1 | 1 - 2 | 395 | 55 | |
Set Membership: |
|