Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965287.1 | XP_017820776.1 | 3 - 4 | 866 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM45A| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965287.1 | XP_017820776.1 | 3 - 4 | 866 | 88 | |
Set Membership: |
|