Assay Details
| Gene Symbol: LOC100405850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763186.3 | XP_002763232.1 | 9 - 10 | 1421 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100405850| Gene Symbol: LOC100405850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763186.3 | XP_002763232.1 | 9 - 10 | 1421 | 92 | |
Set Membership: |
|