Assay Details
| Gene Symbol: TBC1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999981.2 | XP_008998229.2 | 1 - 2 | 1193 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBC1D2| Gene Symbol: TBC1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999981.2 | XP_008998229.2 | 1 - 2 | 1193 | 59 | |
Set Membership: |
|