Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757284.4 | XP_002757330.1 | 7 - 8 | 4397 | 100 | |
| XM_008979893.2 | XP_008978141.1 | 7 - 8 | 3994 | 100 | ||
| XM_008979896.2 | XP_008978144.1 | 7 - 8 | 4397 | 100 | ||
| XM_008979897.2 | XP_008978145.1 | 7 - 8 | 4397 | 100 | ||
| XM_008979898.2 | XP_008978146.1 | 4 - 5 | 3875 | 100 | ||
More Information
Set Membership: |
|

ZNF532
Genomic Map