Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746210.3 | XP_002746256.1 | 1 - 2 | 560 | 62 | |
| XM_008993939.2 | XP_008992187.1 | 1 - 2 | 560 | 62 | ||
More Information
Set Membership: |
|

MCUR1
Genomic Map