Assay Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985687.1 | XP_008983935.1 | 7 - 8 | 1307 | 91 | |
| XM_008985688.1 | XP_008983936.1 | 6 - 7 | 1198 | 91 | ||
| XM_008985690.2 | XP_008983938.1 | 5 - 6 | 571 | 91 | ||
| XM_008985692.2 | XP_008983940.1 | 8 - 9 | 1239 | 91 | ||
| XM_008985693.1 | XP_008983941.1 | 6 - 7 | 1101 | 91 | ||
| XM_008985694.2 | XP_008983942.1 | 5 - 6 | 886 | 91 | ||
More Information
Set Membership: |
|

CNST
Genomic Map