Assay Details
| Gene Symbol: LOC100413740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758316.4 | XP_002758362.1 | 9 - 10 | 1268 | 85 | |
| XM_017964890.1 | XP_017820379.1 | 8 - 9 | 1406 | 85 | ||
| XM_017964891.1 | XP_017820380.1 | 9 - 10 | 1313 | 85 | ||
| XM_017964892.1 | XP_017820381.1 | 9 - 10 | 1268 | 85 | ||
More Information
Set Membership: |
|

LOC100413740
Genomic Map