Assay Details
| Gene Symbol: ALAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762916.1 | XP_002762962.1 | 10 - 11 | 1736 | 64 | |
| XM_002762918.4 | XP_002762964.1 | 9 - 10 | 1625 | 64 | ||
| GenBank mRNA | EF214999.1 | - | 2 - 3 | 430 | 64 | |
More Information
Set Membership: |
|

ALAS2
Genomic Map