Assay Details
| Gene Symbol: LOC100406157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981814.2 | XP_008980062.1 | 12 - 13 | 1911 | 71 | |
| GenBank mRNA | EF215204.1 | - | 1 - 2 | 148 | 71 | |
| EF215281.1 | - | 1 - 2 | 152 | 71 | ||
More Information
Set Membership: |
|

LOC100406157
Genomic Map