Assay Details
| Gene Symbol: LOC100406157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981814.2 | XP_008980062.1 | 13 - 14 | 2000 | 52 | |
| GenBank mRNA | EF215204.1 | - | 2 - 3 | 237 | 52 | |
| EF215281.1 | - | 2 - 3 | 241 | 52 | ||
More Information
Set Membership: |
|

LOC100406157
Genomic Map