Assay Details
| Gene Symbol: DEDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762188.4 | XP_002762234.2 | 2 - 3 | 961 | 59 | |
| XM_008988101.2 | XP_008986349.1 | 2 - 3 | 902 | 59 | ||
| XM_008988102.2 | XP_008986350.1 | 2 - 3 | 999 | 59 | ||
More Information
Set Membership: |
|

DEDD2
Genomic Map