Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755586.4 | XP_002755632.1 | 3 - 4 | 607 | 69 | |
| XM_009008568.2 | XP_009006816.1 | 3 - 4 | 609 | 69 | ||
| XM_017978318.1 | XP_017833807.1 | 2 - 3 | 427 | 69 | ||
More Information
Set Membership: |
|

AIP
Genomic Map