Assay Details
| Gene Symbol: GABRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008993407.2 | XP_008991655.1 | 5 - 6 | 813 | 76 | |
| XM_017970585.1 | XP_017826074.1 | 5 - 6 | 820 | 76 | ||
| XM_017970586.1 | XP_017826075.1 | 2 - 3 | 974 | 76 | ||
More Information
Set Membership: |
|

GABRA2
Genomic Map