Assay Details
| Gene Symbol: EBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762835.4 | XP_002762881.1 | 2 - 3 | 1902 | 68 | |
| XM_008989231.2 | XP_008987479.1 | 3 - 4 | 2138 | 68 | ||
| XM_008989232.2 | XP_008987480.1 | 2 - 3 | 467 | 68 | ||
| XM_008989233.2 | XP_008987481.1 | 3 - 4 | 2091 | 68 | ||
| XM_008989234.2 | XP_008987482.1 | 2 - 3 | 1855 | 68 | ||
| XM_017968574.1 | XP_017824063.1 | 2 - 3 | 414 | 68 | ||
More Information
Set Membership: |
|

EBP
Genomic Map