Assay Details
| Gene Symbol: SLAMF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302667.1 | NP_001289596.1 | 2 | 161 | 63 | |
| XM_008984706.2 | XP_008982954.2 | 2 | 887 | 63 | ||
| XM_017966010.1 | XP_017821499.1 | 2 | 3853 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017966010.1 transcripts.
Set Membership: |
|

SLAMF9
Genomic Map