Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744962.4 | XP_002745008.1 | 5 - 6 | 1189 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SETD9| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744962.4 | XP_002745008.1 | 5 - 6 | 1189 | 63 | |
Set Membership: |
|