Assay Details
| Gene Symbol: SOWAHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980255.2 | XP_008978503.1 | 1 | 475 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOWAHC| Gene Symbol: SOWAHC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008980255.2 | XP_008978503.1 | 1 | 475 | 133 | |
Set Membership: |
|