Assay Details
| Gene Symbol: TNFAIP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744670.4 | XP_002744716.1 | 2 | 1810 | 61 | |
| XM_008991635.2 | XP_008989883.1 | 2 | 5778 | 61 | ||
| XM_008991636.1 | XP_008989884.1 | 2 | 617 | 61 | ||
| XM_008991637.2 | XP_008989885.1 | 2 | 5571 | 61 | ||
More Information
Set Membership: |
|

TNFAIP8
Genomic Map