Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744656.4 | XP_002744702.1 | 1 | 150 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002744656.4 transcripts.
Set Membership: |
|

PRR16
Genomic Map