Assay Details
| Gene Symbol: IL17D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748883.4 | XP_002748929.2 | 1 - 2 | 594 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL17D| Gene Symbol: IL17D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748883.4 | XP_002748929.2 | 1 - 2 | 594 | 65 | |
Set Membership: |
|