Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746030.3 | XP_002746076.1 | 7 - 8 | 1162 | 86 | |
| XM_017970703.1 | XP_017826192.1 | 7 - 8 | 1258 | 86 | ||
| GenBank mRNA | EF215109.1 | - | 2 - 3 | 258 | 86 | |
More Information
Set Membership: |
|

LRPAP1
Genomic Map