Assay Details
| Gene Symbol: LRP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983375.2 | XP_008981623.1 | 5 - 6 | 1776 | 69 | |
| XM_017965639.1 | XP_017821128.1 | 5 - 6 | 1740 | 69 | ||
| XM_017965640.1 | XP_017821129.1 | 6 - 7 | 1635 | 69 | ||
More Information
Set Membership: |
|

LRP12
Genomic Map