Assay Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735620.3 | XP_003735668.1 | 4 - 5 | 623 | 72 | |
| XM_017967795.1 | XP_017823284.1 | 4 - 5 | 447 | 72 | ||
| XM_017967796.1 | XP_017823285.1 | 3 - 4 | 425 | 72 | ||
More Information
Set Membership: |
|

EMP3
Genomic Map