Assay Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968138.1 | XP_017823627.1 | 1 - 2 | 124 | 77 | |
| XM_017968143.1 | XP_017823632.1 | 1 - 2 | 124 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017968138.1, XM_017968143.1 transcripts.
Set Membership: |
|

ZNF529
Genomic Map