Assay Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987822.2 | XP_008986070.1 | 2 - 3 | 212 | 76 | |
| XM_017968141.1 | XP_017823630.1 | 2 - 3 | 153 | 76 | ||
| XM_017968143.1 | XP_017823632.1 | 2 - 3 | 184 | 76 | ||
| XM_017968144.1 | XP_017823633.1 | 2 - 3 | 219 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008987822.2, XM_017968141.1, XM_017968143.1, XM_017968144.1 transcripts.
Set Membership: |
|

ZNF529
Genomic Map