Assay Details
| Gene Symbol: ZNF529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987821.2 | XP_008986069.1 | 5 - 6 | 421 | 118 | |
| XM_008987822.2 | XP_008986070.1 | 4 - 5 | 435 | 118 | ||
| XM_017968137.1 | XP_017823626.1 | 4 - 5 | 2266 | 118 | ||
| XM_017968138.1 | XP_017823627.1 | 5 - 6 | 452 | 118 | ||
| XM_017968139.1 | XP_017823628.1 | 5 - 6 | 487 | 118 | ||
| XM_017968140.1 | XP_017823629.1 | 5 - 6 | 455 | 118 | ||
| XM_017968141.1 | XP_017823630.1 | 4 - 5 | 376 | 118 | ||
| XM_017968143.1 | XP_017823632.1 | 4 - 5 | 407 | 118 | ||
| XM_017968144.1 | XP_017823633.1 | 4 - 5 | 442 | 118 | ||
More Information
Set Membership: |
|

ZNF529
Genomic Map