Assay Details
| Gene Symbol: MUC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965834.1 | XP_017821323.1 | 6 - 7 | 1561 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MUC20| Gene Symbol: MUC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965834.1 | XP_017821323.1 | 6 - 7 | 1561 | 62 | |
Set Membership: |
|